VWR® Glass Disposal Box
# de catalogue 470204-582
Fournisseur: VWR International
Spécifications
- Dimensions:8×8×10"
- Description:VWR® Glass Disposal Box, Bench Top
- Cat. no.:470204-582
Spécifications
A propos de cet article
These handy disposal boxes keep broken glass contained and maximize safety in your lab or classroom.
- Durable, Corrugated Cardboard
- Polypropylene Bag Included
- Quick and Easy Cleanup
- For Broken Glass and Spilled Liquids
Each receptacle includes a 2 mm thick polypropylene bag sturdy enough to hold broken glass fragments and spilled liquids.
Pull the safety cap into place when the box is full for easy clean up and dispose of the entire unit.
Customers Who Bought This Also Bought