VWR® Starter Glassware Set
# de catalogue 470103-580
Fournisseur: Avantor
Spécifications
- Description:VWR® Starter Glassware Set
- Cat. no.:470103-580
- Supplier No.:WARD470103-580
Spécifications
A propos de cet article
VWR® Starter Glassware Set - Set includes one 100 ml, one 250 ml, and one 400 ml beaker, 12 pk of droppers, one 10 ml, one 50 ml, and one 10 0ml Graduated Cylinder, one 65 mm long stem funnel, six 13x100 mm test tubes, six 18x150 mm test tubes, and six 25x150 mm test tubes, along with a plastic test tube rack.
Customers Who Bought This Also Bought