Ward's Compact Scales
Catalog # 470314-390
Supplier: OHAUS CORP.
Specifications
- Pan/Platform Size:142×128 mm
- Dimensions:135W×41H×201D mm
- Repeatabilty:1.0 g
- Cat. no.:470314-390
- Readability:0.1 g
- Weighing Capacity:1200 g
Specifications
About this item
These portable, compact scales are an economic option for the lab.
- Auto shut-off
- One-year manufacturer's warranty
- High-contrast LCD Screen
- Two-button keypad
- Stackable
- Aligned with GLOBE protocols
These models feature a simple two-button keypad and high-contrast LCD screen for ease of use and visibility.
BodyTextAccessories: An AC adapter is available separately.
Ordering information: One-year manufacturer's warranty is included with purchase. The scales include three AA batteries.
Customers Who Bought This Also Bought