VSEPR SuperModel Kit
# de catalogue 470002-700
Fournisseur: RYLER ENTERPRISES, INC.
Spécifications
- Description:VSEPR SuperModel Kit
- Cat. no.:470002-700
- Supplier No.:VSP1
Spécifications
A propos de cet article
Model the Theory of Valence Shell Electron Pair Repulsion
- Includes instruction booklet with activities
- Build all types of atomic structures
Students will learn about molecular geometry, resonance, and molecular polarity as they work through the activity in the included instruction booklet. They will utilize the octet rule to create linear, tetrahedral, pyramidal, and bent trigonal planar models. The set contains seventeen colored atoms and sixteen clear bonds, as well as various spare parts.
Customers Who Bought This Also Bought