Ward's® Voltage and Currents Lab Activity
# de catalogue 470013-324
Fournisseur: Science First
Spécifications
- Description:WARD’S Voltage and Currents Lab Activity
- Cat. no.:470013-324
- Supplier No.:14476
Spécifications
A propos de cet article
Explore Direct Current Circuits
Examine the effects varying voltage has on common components in simple, DC circuits. See how a standard 6 V light bulb varies in brightness, temperature, and color at different voltages, and the changes in intensity and blinking rate in a 2.5 V “blinky” bulb. You can also see how voltage and circuit polarity effect the operation and intensity of an LED. An optional investigation introduces circuit diagrams and the relationship between current and voltage. It includes enough bulbs, connectors, LEDs, and more for five setups, as well as a teacher’s guide and student instructions. Four AA batteries per setup, required, are available separately.
Customers Who Bought This Also Bought