Volta's Hail Storm
Fournisseur: Avantor
Certificats
A propos de cet article
A Perfect accessory for the Electrostatic Generator
- Simple to use
- Attaches to any Van De Graaff
Place pith balls or bits of aluminum foil in this cylinder to see how particles are attracted to the upper plate by induction, then repelled upon contact.
Customers Who Bought This Also Bought