Vial Labels for Drosophila Crosses,
# de catalogue 470022-582
Fournisseur: TLF GRAPHICS
Spécifications
- Description:Vial labels for drosophila crosses
- Cat. no.:470022-582
- Supplier No.:320291
Spécifications
A propos de cet article
Pressure-sensitive labels for recording parents of Drosophila crosses.
The markings won’t rub off.
Packaging: Roll of 500.
Customers Who Bought This Also Bought