Vascular System Model
# de catalogue 470029-544
Fournisseur: KYOTO KAGAKU AMERICA, INC.
Spécifications
- Description:Vascular System
- No. of Keyed Items:240
- Cat. no.:470029-544
- Supplier No.:A61
- No. of Parts:1
Spécifications
A propos de cet article
A Unique Model Illustrating Blood Flow.
- 3/4 Size
- Hardened Metal Construction
- Includes Key
It’s the perfect way to acquaint students with the way blood flows through a human body. Simulating a corrosion preparation, only the Vascular system is visible. The arterial system is shown in red, the venous system in blue. The 3/4 life-size model is constructed of hardened plastic-coated metal wire, is supported by a metal rod. The model is mounted on a metal base and includes a key identifying 240 structures.
Customers Who Bought This Also Bought