ContactUsLinkComponent
Variable Inductor
Variable Inductor
# de catalogue 470128-394
Fournisseur:  VWR International
INDUCTOR VARIABLE
Variable Inductor
# de catalogue 470128-394
Fournisseur:  VWR International
Spécifications
  • Description:
    Variable Inductor
  • Cat. no.:
    470128-394
  • Supplier No.:
    PH0845VIC
Spécifications
A propos de cet article
Adjustable for Precision
  • Compatible with 4 mm safety sockets.
  • Includes a carrying handle for easy portability. 
The screw adjustment enameled wire winding built into this unit allows precise measurements up to 1.3 H. Soft iron core displacement includes a scale mark and quick advancement worm.
Customers Who Bought This Also Bought