Kemtec® Urinalysis
# de catalogue 470111-836
Fournisseur: Kemtec
Spécifications
- Description:Urinalysis Kit
- Cat. no.:470111-836
- Supplier No.:8-306
Spécifications
A propos de cet article
Simulated urine testing.
- No real or human products
- Learn basic testing procedure
Urinalysis introduces students to to the world of biomedical technology. Using simulated samples, students evaluate the general characteristics of urine, observe for sediments, and test for glucose, albumin, pH, specific gravity, and lead.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought