Ward's® Ultramafic Rocks Thin Sections Set
# de catalogue 470024-960
Fournisseur: Avantor
Spécifications
- No. of Slides:12
- Description:WARD’S Ultramafic Rocks Thin Section Set
- Cat. no.:470024-960
- Supplier No.:WARD470024-960
Spécifications
A propos de cet article
Enhance Your Ultramafic Studies with This Special Petrographic Resource
- Hand Made by Wards® staff Geologists
- 12 Specially Selected Samples
- Microscopic view of Igneous Samples with Very Low Silica Content
Due to the growing demand and interest in ultramafic rocks, we’ve prepared a set of twelve thin sections to match our Ultramafic Rock Collection. Each slide has been selected to illustrate the key textural and compositional features of ultramafic rocks.
Contents: bronzitite, carbonatite, dunite (2), kimberlite, lamproite, peridotite (3), pyroxenite, serpentinite, soapstone
Customers Who Bought This Also Bought