Types of Fossilization Collection
# de catalogue 470111-000
Fournisseur: SCOTT RESOURCES 2, LLC SE
Spécifications
- Description:Types of Fossilization Collection
- Cat. no.:470111-000
- Supplier No.:3010-S
- No. of Parts:16 samples
Spécifications
A propos de cet article
This Collection has been assembled to provide background into the history of life on Earth.
- Collection Contains Real Fossils Collected from Multiple Locations
- All Major Methods of Fossilization Represented
- Identification Key Included
- Average Sample Size: 1/2 to 1"
- Fifteen Fossils Included
Each set contains individually numbered specimens housed in a compartmented collection box with removable tray and key list. The average sample size is 1/2 to 1".
This illustrative set of 15 fossils displays examples of the primary forms of fossilization including direct replacement (pyritization, calcification, silification), molds and casts, distillation (carbonization), and unaltered remains.
Customers Who Bought This Also Bought