High-Vacuum Pumps
Fournisseur: Fischer Technical
Certificats
A propos de cet article
High-Vacuum Pumps are ideal for demonstrations and lab work.
- Intake Connections Via 3/8" ID Tubing
- Pressure Low Enough for Boyle's Law Experiments
- Version with Gauge Reduces Errors Due to Poor Seals
- CSA Approved
- Fitting for Demos and Lab Work
Intake connections are via 3/8" ID tubing. Both models deliver pressure low enough for all Boyle's Law experiments.
The version with gauge reduces errors due to a poor seal between the pump and the vacuum plate.
Airflow: 3 cfm
Max vacuum: 25 microns (0.025 mm of Hg)
120 V
Certifications: CSA approved.
Customers Who Bought This Also Bought