Fruit and Potato Clock
# de catalogue 470006-220
Fournisseur: VWR International
Spécifications
- Description:Two Potato Clock
- Cat. no.:470006-220
- Supplier No.:PT-CLK
Spécifications
A propos de cet article
This LCD clock keeps time using potatoes, soda, or plants.
- Length: 8½"
- Width: 3½"
- Height: 2¾"
- Demonstration of an Electrochemical Cell
- Instructions Included
It's an ideal demonstration of an electrochemical cell. All you need to do is attach the included zinc and copper bimetallic probes and watch the resettable hours, minutes, seconds, and date light up.
The clock includes instructions and measures 2¾ x 3½ x 8½".
Customers Who Bought This Also Bought