Tuning Fork and Resonator Box Set
# de catalogue 470111-232
Fournisseur: United Scientific Supplies
Spécifications
- Description:Tuning Fork and Resonator Box Set
- Cat. no.:470111-232
- Supplier No.:TFRB256
Spécifications
A propos de cet article
Produces a Louder Tone With a Greater Range of Audible Overtones
- Tuning fork is securely mounted on top
- Open end box increase the sound of the resonance
A physical pitch tuning fork has been specifically modified to screw into a resonator case. The unit consists of a tuning fork, wooden resonator box and a wooden wedge snapper for exciting the fork.
Customers Who Bought This Also Bought