Tuning Fork Activator
# de catalogue 470148-966
Fournisseur: VWR International
Spécifications
- Description:Tuning Fork Activator
- Size:10 x 7.6 x 5 cm
- Cat. no.:470148-966
- Supplier No.:P36142
Spécifications
A propos de cet article
The activator is specifically designed for use with tuning forks.
- Durable
- Reliable
- Rubber Construction
- Activator Prevents Damage
- For Use with Tuning Forks
Striking delicate forks against each other can cause nicks and effect the tuning. This activator prevents that damage.
With solid rubber construction, it's durable and reliable.
Customers Who Bought This Also Bought