Tubby® Storage Container
Catalog # 470105-518
Supplier: Heathrow Scientific
Specifications
- Description:Tubby® storage container
- Dimensions:33×20×11.5 cm (13×7.9×4.5")
- Cat. no.:470105-518
- Supplier No.:HS23453
Specifications
About this item
Tubby® stackable storage container is designed to help organize a laboratory.
- The lid snaps in place to secure the contents and for safe transport
- Constructed of high-impact polypropylene
The removable dividers can create as big or as small a compartment as needed. Perfect for keeping tubes, tips, pipettes, gloves and other small items organized and out of the way.
Customers Who Bought This Also Bought