Tryptic Soy Broth
# de catalogue 470015-844
Fournisseur: Avantor
Numéro CAS:
Spécifications
- Description:Dehydrated Tryptic Soy Broth
- Cat. no.:470015-844
- Supplier No.:WARD470015-844
Spécifications
A propos de cet article
Soybean-casein digest medium. Supports the growth of a variety of microorganisms.
Premeasured Packs Save Time in the Lab
Simply add the contents of one Dehydrated Media pack to one liter of water, bring to a boil, and sterilize, to make enough media for 40 to 50 plates or 16edited0 tubes.
Customers Who Bought This Also Bought