Prepared Tryptic Soy Broth Media
Fournisseur: Culture Media & Supplies
Certificats
A propos de cet article
This is a good all-purpose medium that supports the growth of a variety of microorganisms.
- All-purpose media
- Pre-prepared, just inoculate and incubate
This nutrient rich broth is a non-selective media that is used to grow most types of aerobic organisms. Simply pour into sterile Petri dish plates.
If you are looking for a similar media, but in solid agar form, please select Tryptic Soy Agar (available in prepared plates, tubes, and as a dehydrated, ready to-mix powder)
Caution: These products are not lot controlled and are for educational use only.
Customers Who Bought This Also Bought