Trypic Soy Agar, Dehydrated
# de catalogue 470193-226
Fournisseur: IPM SCIENTIFIC, INC.
Numéro CAS:
Spécifications
- Description:Dehydrated Tryptic Soy Agar
- Cat. no.:470193-226
- Supplier No.:880002
Spécifications
A propos de cet article
Soybean-casein digest medium supports the growth of a variety of microorganisms. Premeasured packs save time in the lab. Simply add the contents of one dehydrated media pack to one liter of water, bring to a boil and sterilize to make enough media for 40 to 50 plates.
Customers Who Bought This Also Bought