Prepared Tryptic Soy Agar Media
Fournisseur: Culture Media & Supplies
Certificats
A propos de cet article
This soybean-casein digest medium can support a wide variety of microorganisms.
- Sterile, pre-prepared media; just inoculate and incubate.
- Excellent, basic medium that supports most microorganisms.
This medium is an excellent choice for general subculture for storage, colony counts as well as streak-plating for isolates.
Soft tube package contains: 12 tubes of tryptic soy agar for use in plaque morphology studies.
Pour-pack includes: 3 bottles of prepared Tryptic Soy Agar and 20 sterile Petri dishes.
Designed for educational and teaching laboratories - no COA/COQ is available
Customers Who Bought This Also Bought