Trolley Picket Fence
Catalog # 470134-170
Supplier: VWR International
Specifications
- Description:Trolley Picket Fence
- Cat. no.:470134-170
- Supplier No.:PIC-FNC
Specifications
About this item
Study Acceleration and Velocity without Cart or Track
- Recommended for use with photogates
All you need is a string and a set of photogates to create the perfect way to study acceleration and velocity without a dynamics cart or track. A photogate on support stands allows for the adjustment of the picket fence so it slides through the photobeam.
Customers Who Bought This Also Bought