Tris-Glycine Buffer
# de catalogue 470120-920
Fournisseur: Edvotek
Numéro CAS:
Spécifications
- Description:Tris-Glycine Buffer
- Size:500 mL
- Chemical/Physical Specifications:10X Solution
- Cat. no.:470120-920
- Grade:Laboratory
- Supplier No.:656
Spécifications
A propos de cet article
Running buffer for native PAGE.
Caution: Please Note: This product is designed for educational and teaching laboratories, and no certificate of analysis is available.
Customers Who Bought This Also Bought