Tri-fold Field Shovel
# de catalogue 470013-336
Fournisseur: LIBERTY MOUNTAIN SPORTS, LLC
Spécifications
- Length:23"
- Cat. no.:470013-336
- Weight:36 oz.
- Supplier No.:S031
Spécifications
A propos de cet article
This compact and lightweight folding shovel can be used at a 90° angle for chopping or straight for shovelling, or it can be securely locked in three positions to provide the best angle for the job. The blade also has a serrated edge for cutting roots. Folded 10".
- Aligned with GLOBE protocols
Customers Who Bought This Also Bought