Tree Identification Kit
# de catalogue 470102-342
Fournisseur: Young Naturalist
Spécifications
- Description:Tree Identification Kit
- Cat. no.:470102-342
- Supplier No.:301
Spécifications
A propos de cet article
Students learn how to key the 13 different leaves including elm, ash, walnut, maple, pine, cedar, locust, oak, cottonwood, and willow.
- Each actual leaf specimen is individually bagged
- Students can then use the included Tree Finder book, also sold separately, to identify their local trees
Includes leaf key and materials for 24 students.
Customers Who Bought This Also Bought