Translucent Plastic Case
Catalog # 470157-552
Supplier: DR INSTRUMENTS INC. SE
Specifications
- Description:Translucent Plastic Case
- Cat. no.:470157-552
- Supplier No.:505P
Specifications
About this item
Organize your dissection tools in this clear plastic case, so you can easily see what instruments are available without sorting through its contents.
The autoclavable case features individual compartments for every tool, and a hinged top. Size: 3"W x 6.75"L x 0.75"H.
Customers Who Bought This Also Bought