ContactUsLinkComponent
Ward's® Transduction of E.coli Lab Activity
Ward's® Transduction of E.coli Lab Activity
# de catalogue 470024-178
Fournisseur:  Avantor
WARD'S TRANSDUCTION/E COLI 15 GROUPS
Ward's® Transduction of E.coli Lab Activity
# de catalogue 470024-178
Fournisseur:  Avantor
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Transduction of E. coli Kit
  • No. of Lab Groups:
    8
  • Cat. no.:
    470024-178
Spécifications
A propos de cet article
Use Viruses to Deliver Genetic Material into Bacteria
  • Discuss applications for viral-induced genetic engineering
  • Materials for 8 lab groups
  • Time Required: ~120 minutes
Demonstrate transduction using a mutant E. coli, which is incapable of producing beta-galactosidase for lactose breakdown, and a lambda phage carrying the lac+ phenotype. Students will observe and differentiate between plaques and colonies to discover how this relates the lytic and lysogenic phases of transduction, and then will use their results to calculate the incidence of transduction for each plate. It includes enough materials for eight setups, a teacher’s guide, and student copymaster.
Ordering information: Coupon is included for redemption of perishable materials. Redeem by mail, phone, fax, or email. Activity includes 2 Tubes of tryptic soy broth; 2 Bottles; Luria agar; 1 Sleeve; Petri plates; 2 Cotton swabs; 2 Vials of N,N-dimethylformamide; 50 Disposable pipettes; 16 Capped vial; 20 Bacti-spreaders; 1 Tube of Sterile water; 1 Redemption coupon for: 1 Culture of E. coli host strain; 1 Tube of lambda phage gt11; 2 Vials of X-gal; and 2 Vials of IPTG.
Customers Who Bought This Also Bought