Thistle Tube Polypropylene Funnel, Bel-Art
# de catalogue 470177-142
Fournisseur: Bel-Art
Spécifications
- Description:Thistle Tube Polypropylene Funnel
- Cat. no.:470177-142
- Supplier No.:F147510000
Spécifications
A propos de cet article
This unbreakable plastic thistle tube is ideal for the high school laboratory. It has a two piece construction with the stem tightly fitting into the funnel. Stem O.D.: 6mm. Length: 290mm. Package of 12.
Customers Who Bought This Also Bought