Workstation Organizer
# de catalogue 470008-092
Fournisseur: Heathrow Scientific
Spécifications
- Description:Workstarion organizer
- Cat. no.:470008-092
- Supplier No.:HS2538A
Spécifications
A propos de cet article
Polystyrene organizer is perfect for storing and organizing small laboratory supplies.
- Protect small and fragile items
- Fit in most standard drawers or store on benches and shelves
Pipettes, thermometers, and other long, breakable items have a safe storage spot.
Three plastic compartments, two small 17×3.1" and one large 17×4.3", keep instruments flat and separated. Overall size: 17.9×12.1×2.5".
Customers Who Bought This Also Bought