Thermometer/Pipette Tray
# de catalogue 470145-364
Fournisseur: Bel-Art
Spécifications
- Description:Thermometer/Pipette Tray, Each
- Cat. no.:470145-364
- Supplier No.:H18648-0000
Spécifications
A propos de cet article
Molded of high impact plastic to fit into laboratory bench drawers, this tray has three compartments for storage of thermometers or pipettes.
Tray dimensions: 14" x 17" and 2-1/2" deep.
Customers Who Bought This Also Bought