Thermo-Anesthetizer Set
# de catalogue 470007-112
Fournisseur: Avantor
Spécifications
- Description:Thermo-anesthetizer set
- Cat. no.:470007-112
- Supplier No.:WARD470007-112
Spécifications
A propos de cet article
These reusable “bricks”, with their own covered Petri dishes, store in the freezer until ready for use. They immobilize Drosophila safely without chemical usage.
- Flies on the plate are immobilized for up to 20 minutes
- Each brick may be reused several times
Brick size W×L×H: 4½×4×1½"
Ordering information: Package of 24 bricks and Petri dishes.
Delivery: WARD’S Drosophila Culture Guide is included.
Customers Who Bought This Also Bought