The Stirling Design External Combustion Engine
# de catalogue 470008-038
Fournisseur: PM RESEARCH INC.
Spécifications
- Description:Stirling Engine, External Combustion, 18 cm x 11 cm
- Cat. no.:470008-038
- Supplier No.:SOLAR 1 W/
Spécifications
A propos de cet article
Demonstrate the Stirling Design External Combustion Engine with our 18×11 cm working model. The Stirling engine currently is being re-examined as a practical source of power because of its adaptability to almost any fuel. First produced in the 1800’s as a non-electric fan and an automatic hot air water pump, the engine soon became obsolete due to its lack of sufficient power when compared to the newly developed combustion engine. Any fuel can be used with our model, but denatured alcohol is suggested for clean operation.
The book Stirling Cycle Engines by Andy Ross and an owner's manual are included. Due to the nature of this item, it cannot be shipped by air under any circumstances. Please select ground shipping at time of checkout.
Customers Who Bought This Also Bought