The Remagnetizer
# de catalogue 470003-722
Fournisseur: Master Magnetics
Spécifications
- Description:The Remagnetizer
- Cat. no.:470003-722
- Supplier No.:DM-RM
Spécifications
A propos de cet article
Lightweight and reliable
- Instructions are included
- CSA approved
- Size: 10 x 13 x 7cm
Save yourself time and money by magnetizing your Alnico horseshoe and bar magnets when they become weak. This handy tool contains two permanent neodymium magnets to simply and quickly recharge your magnets. The Remagnetizer can be mounted to a wall or table and requires no electricity. It has a durable aluminium frame and steel base, and weighs only 1.38 kg (3.05 lbs.).
Customers Who Bought This Also Bought