Ward's® The Cell Theory: A Microscopic Journey Lab Activity
# de catalogue 470012-252
Fournisseur: Avantor
Spécifications
- Description:The Cell Theory: A Microscopic Journey Lab Activity
- No. of Lab Groups:8
- Cat. no.:470012-252
- Supplier No.:WARD470012-252
Spécifications
A propos de cet article
Unique Approach To Study Cellular Structures
- Teacher Developed
- Pre And Post-Laboratory Assessments
- Links To Real World Concepts
- Estimated Class/Lab Time Required: 45 Minutes
Travel back in time for an exploration of scientific history. This laboratory leads students through the key discoveries of noted scientists during the one hundred year development of the Cell Theory. Each of the provided prepared slides helps students see what the leading researchers of cell theory saw in their studies.
Materials Included:
Slides
Slide holder
Biography of each scientist
Materials Needed But Not Supplied: Microscopes
Customers Who Bought This Also Bought