The Brain Poster
# de catalogue 470112-294
Fournisseur: Anatomical Chart Co.
Spécifications
- Length:66 cm
- Width:50 cm
- Description:Brain Poster
- Cat. no.:470112-294
- Finish:Laminated
- Supplier No.:1587791056
Spécifications
A propos de cet article
- Eleven full color illustrations
Poster shows lobes of the brain, blood vessels, origin of cranial nerves, areas of cortex, ventricles, circle of Willis and other related structures.
Customers Who Bought This Also Bought