The Atlantis Ocean Globe
# de catalogue 470006-238
Fournisseur: Replogle Globes
Spécifications
- Description:The Atlantis Ocean Globe
- Cat. no.:470006-238
- Supplier No.:33801
- Diameter:30.5 cm
Spécifications
A propos de cet article
The globe is mounted in a gold-finished, fully numbered meridian ring, which swings either up or down for easy viewing.
- Shows submarine physical features
- Topography is shown in raised relief
Nicely illustrated, this world globe shows the physical features under the sea, as well as mountains, deserts, grasslands and forests in raised relief. Size: 30.5cm dia.
Customers Who Bought This Also Bought