Tetrahymena Medium
# de catalogue 470180-756
Fournisseur: Avantor
Numéro CAS:
Spécifications
- Description:Tetrahymena Medium
- Cat. no.:470180-756
- Supplier No.:WARD470180-756
Spécifications
A propos de cet article
Sterile. For culture of Tetrahymena. Supplied in 20 x 150 mm tubes. Package of 12.
Customers Who Bought This Also Bought