Wood Test Tube Rack
# de catalogue 470104-542
Fournisseur: Bennett Wood Specialties
Spécifications
- Description:Wood Test Tube Rack
- Cat. no.:470104-542
- Supplier No.:6327900
Spécifications
A propos de cet article
One-piece support is able to accommodate larger size tubes. It holds six 20mm tubes. Size: 12 x 6 x 4.5cm.
Customers Who Bought This Also Bought