Test Lead Wire Set
# de catalogue 470122-156
Fournisseur: Electric Switches
Spécifications
- Description:Test lead wire set
- Cat. no.:470122-156
- Supplier No.:110003
Spécifications
A propos de cet article
Set contains one red and one black 48" test lead with right angle banana plugs, two test lead insulators — one red, one black, two plunger-type clips — one red, one black and one red spade lug, two clip insulators — one red, one black, and alligator clips.
Customers Who Bought This Also Bought