Ward's® Terrain Feature Model
# de catalogue 470029-382
Fournisseur: Plas-Labs
Spécifications
- Description:WARD’S Terrain Feature Model
- Cat. no.:470029-382
- Supplier No.:151-TM
Spécifications
A propos de cet article
Expand the Number of Activities You Can Perform with Your Stream Table
Add the plastic mountain region to the lake area of your stream table to study the movement of water and sediment in relation to a landform; flood the lake region at regular depth increments, then develop a contour map of your observations and measurements. Size: 15"L x 11 1/2"W x 3"H.
Customers Who Bought This Also Bought