ContactUsLinkComponent
Ten Specimen Density Set
Ten Specimen Density Set
# de catalogue 470149-490
Fournisseur:  VWR International
DENSITY SET - 10 SPECIMENS
Ten Specimen Density Set
# de catalogue 470149-490
Fournisseur:  VWR International
Spécifications
  • Description:
    Ten Specimen Density Set
  • Cat. no.:
    470149-490
  • Supplier No.:
    P10040
Spécifications
A propos de cet article
This set features a handy, economical selection of density cubes.
  • Economical
  • Handy
  • Cubes Measure 1" Each
  • Materials Include Pine, Poplar, Plastic, and More
  • Ten Density Cubes per Set
The cubes measure 1" each.
They're made of acrylic plastic, aluminum, brass, copper, nylon, oak, pine, poplar, PVC, and steel.
Customers Who Bought This Also Bought