Tangent Height Gauge
# de catalogue 470018-570
Fournisseur: Peco Sales
Spécifications
- Description:Tangent Height Gauge
- Cat. no.:470018-570
- Supplier No.:36953
Spécifications
A propos de cet article
Teach Students to Quickly Determine Tree Heights
This mirror sighting gauge includes easy to follow instructions embossed directly onto its rugged plastic shell. The instructions detail the gauge’s short four step measuring process as well as height-finding formulas.
Customers Who Bought This Also Bought