ContactUsLinkComponent
Kemtec® Tallquist Hemoglobin Scale
Kemtec® Tallquist Hemoglobin Scale
# de catalogue 470023-872
Fournisseur:  Kemtec
Kemtec® Tallquist Hemoglobin Scale
# de catalogue 470023-872
Fournisseur:  Kemtec
Restricted Products: To process your orders without delay, please provide the required business documentation to purchase this product.
To order chemicals, medical devices, or other restricted products please provide ID that includes your business name & shipping address via email [email protected] or fax 484.881.5997 referencing your VWR account number. Acceptable forms of ID are:
  • • State issued document with your organization's Federal Tax ID Number
  • • State issued document with your organization's Resale Tax ID Number
  • • City or County issued Business License
  • • State Department of Health Services License
  • • Any other ID issued by the State that includes the business name & address
* ATTN: California Customers may require additional documentation as part of the CA Health & Safety Code. Products that fall under this regulation will be placed on a mandatory 21-day hold after documentation is received. VWR will not lift restrictions for residential shipping addresses.
Spécifications
  • Description:
    Tallquist Hemoglobin Scale
  • Cat. no.:
    470023-872
  • Supplier No.:
    12-136
Spécifications
A propos de cet article
Standard hemoglobin reference scale.
  • A fast test to determine anemia levels
  • Paper for 150 tests
The Tallquist hemoglobin scale allows a quick assessment of a patient's hemoglobin levels. Allow blood to absorb into one of the test papers and compare the blood to the color scale to determine the concentration of hemoglobin in blood under normal and anemic conditions.
Delivery: See datasheet for contents information.
Customers Who Bought This Also Bought