ContactUsLinkComponent
Tackling Science Kit: The Basics of Acids and Bases
Tackling Science Kit: The Basics of Acids and Bases
# de catalogue 470152-936
Fournisseur:  Avantor
KIT TACKLING SCIENCE: ACIDS AND BASES
Tackling Science Kit: The Basics of Acids and Bases
# de catalogue 470152-936
Fournisseur:  Avantor
Spécifications
  • Description:
    Tackling Science Kit: The Basics of Acids and Bases
  • Cat. no.:
    470152-936
  • Supplier No.:
    WARD470152-936
Spécifications
A propos de cet article
Tackling Science: Hooking your Kids on Science
  • Enhance your current curriculum
  • Includes all activity supplies
Enhance your current science curriculum with these innovative lesson supplements. Each kit of materials contains everything you need to tackle a variety of hands-on activities focusing on a specific subject of science study. Easily integrated into any program, these inquiry-based learning kits are ideal for science centers, enrichment, or take home explorations. Developed by Teresa Harris Matherne, all Tackling Science Kits include a student science journal, reusuable laminated student guide, and all activity supplies packaged in a stackable tackle box. For grades 2-4.
Litmus paper is used as a tool, students tests baking soda, vinegar, and other liquids to explore what it means to be an acid, base, or neutral substance.
Customers Who Bought This Also Bought