Suture Training Kit
# de catalogue 470128-036
Fournisseur: DR INSTRUMENTS INC. SE
Spécifications
- Description:Suture Training Kit
- No. of Lab Groups:1
- Cat. no.:470128-036
- Supplier No.:SK10
Spécifications
A propos de cet article
All-In-One Training Kit.
- Convenient Carrying Case
- Tool and Practice Pad Storage
- Large Multilayer Practice Pad
The kit includes a large, three-layer skin pad and secure Velcro fasteners with six suture instruments for repeated practice in a variety of suture techniques.
The equipment comes in an attractive and durable carrying case with six instrument loops and a pocket for skin-pad storage. The secure zipper closure and convenient carrying handles make this kit ideal for on-the-go practice in almost any location.
Caution: This kit is for training purposes only and is not for use on humans or animals.
Customers Who Bought This Also Bought