Sun Tracking Hemisphere Kit
# de catalogue 470007-884
Fournisseur: Science First
Spécifications
- Description:Sun Tracking Hemisphere Kit, Set of 10
- No. of Lab Groups:10
- Cat. no.:470007-884
- Supplier No.:PS-03
Spécifications
A propos de cet article
Make Your Students Active Participants in Long-Duration Observations
- Observe changes in the Sun's path over time
Many students believe that the sun is at zenith at noon for every day of the year. By making this model, they will learn the true daily path of the sun. Once the model is created, students will repeat the experiment one month later and then compare the daily paths of the sun for different seasons. Includes ten hemispheres, one pen, and instructions with activities.
Customers Who Bought This Also Bought