Sturdy Covered Cardboard Containers
Catalog # 470095-442
Supplier: RDA CONTAINER CORPORATION SE
Specifications
- Description:Box,Chest,w/FlipTopLid, 24"x12"x10" (60 cm x 30 cm x 25 cm)
- Cat. no.:470095-442
- Supplier No.:2999800
Specifications
About this item
An economical storage solution. All boxes are made of plastic-coated, double-ply cardboard, and feature flip-top lids.
Frequently Bought Together