Student Workstations, Single-Faced Units Phenolic Top, Full Cupboard Configuration
Fournisseur: Diversified Woodcrafts
Certificats
A propos de cet article
Single faced tables that provide a fully equipped science center from two to eight students.
- 3/4" phenolic resin surface
- Constructed of solid oak and oak hardwood veneers
- The cabinets have locking cupboard
- The unit is finished with a chemical resistant, earth-friendly UV finish, rubber base molding and leg boots
The single faced units contain a solid epoxy resin sink along with cold water faucets for every two students. Every unit contains enough gas cocks for each student along with GFI AC duplex electrical receptacles, rod sockets and book compartments. All units contain locks for the drawers and doors. Add 66" in length for each additional two students.
Sink dimensions: 12L×8W×6"H
Certifications: SEFA compliant and MAS Certified Green.
Ordering information: Choose from models with a full cupboard only or a half cupboard and dovetailed drawers. Can be ordered without sink, fixtures and rod sockets. Your choice of 1" epoxy resin or ¾" phenolic resin tops.
Delivery: Four student single faced workstation includes two sinks, four cold water/gas fixtures, two GFI protected AC duplex receptacles, four rod sockets and four book compartments in the aprons.
Spécifications
- Catalog No:
- 470117-662
- 470117-660
- 470117-658
- Environmentally Preferable:
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Safer Human and Environmental Health
- Environmental certification:
- MAS Certified Green
- MAS Certified Green
- MAS Certified Green