Spécifications
- Description:Student Spectrometer
- Cat. no.:470007-298
- Supplier No.:PH0619
Spécifications
A propos de cet article
This spectrometer is economical and capable of performing many types of experiments involving spectras, angle of prisms, refractive indexes, and more.
This spectrometer may be economically priced, but it's still capable of performing many different types of experiments involving spectras, angle of prisms, refractive indexes and more.
This Student Spectrometer consists of a heavy base with a 170mm diameter circle, divided into 360 degrees, which rotates independently and has a locking screw. Other fine features of the spectrometer include: a fixed collimator with an adjustable axis; an objective lens with a quality, lockable focusing system, a variable aperture slit and a Ramsden eyepiece with cross-wires; a moveable telesope featuring a set screw, a fine motion screw and an adjustable axis, and a telescope pillar with a vernier scale for accurate measurements; a prism table with 3 leveling screws and engraved lines and a clamp for prism placement in experiments; and a grating holder.
Comes with an instruction sheet. A monochromatic light source is required but not included. Prism and diffraction gratings not included.
- Variable Aperture Slit
- Eyepiece with Cross-Wires
- Set Screws to Hold Positions
- Economical
- Instruction Sheet Included
This Student Spectrometer consists of a heavy base with a 170 mm diameter circle, divided into 360", which rotates independently and has a locking screw.
Other features of the spectrometer include a fixed collimator with an adjustable axis; an objective lens with a lockable focusing system, a variable aperture slit, and a Ramsden eyepiece with cross-wires; a moveable telescope featuring a set screw, a fine motion screw, an adjustable axis, and a telescope pillar with a vernier scale for accurate measurements; a prism table with three leveling screws, engraved lines, and a clamp for prism placement in experiments; and a grating holder. A monochromatic light source is required but not included.