Student Safe-T Compass
# de catalogue 470089-478
Fournisseur: Learning Resources
Spécifications
- Description:Student Safe-T compass
- Cat. no.:470089-478
- Supplier No.:45759
Spécifications
A propos de cet article
Compass with straight edges made up of shatter-resistant plastic draws circle from 1/4" to 10" in 1/8" diameter increments.
Can be used with pencil or ball-point pen and the straight edge is calibrated inch/metric rules.
Delivery: Supplied with instructions.
Customers Who Bought This Also Bought